Human Recombinant CD147/EMMPRIN/Basigin Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10186-H08H-1
77446-386EA 2258 USD
77446-386
Human Recombinant CD147/EMMPRIN/Basigin Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-His 205) of human CD147 (NP_940991.1) precursor was expressed with the fused polyhistidine tag at the C-terminus.