Human Recombinant IL-1R8/IL1RAPL1 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10177-H02H-1 10177-H02H-100
77446-376EA 348 USD
77446-376 77446-374
Human Recombinant IL-1R8/IL1RAPL1 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human IL1R8 (NP_055086.1) extracellular domain (Met 1-Leu 354) was fused with the Fc region of human IgG1 at the C-terminus.