Human Recombinant CSF1R Protein (Fc & AVI Tag) (from HEK293 Cells), Biotinylated
Supplier: Sino Biological
10161-H41H-B-20 10161-H41H-B-100
77446-360EA 298 USD
77446-360 77446-358
Human Recombinant CSF1R Protein (Fc & AVI Tag) (from HEK293 Cells), Biotinylated
Proteins and Peptides
A DNA sequence encoding the human CSF1R (NP_005202.2)(Met1-Glu512) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.