Human Recombinant Cadherin-8/CDH8 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10144-H08H-50
77446-338EA 248 USD
77446-338
Human Recombinant Cadherin-8/CDH8 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of human CDH8 (NP_001787.2) (Met 1-Met 621) was expressed with a C-terminal polyhistidine tag.