BloodStor® 100 Freezing Medium
Supplier: STEMCELL Technologies
07938 07939 07951
MSPP-07938EA 868.15 USD
MSPP-07938 MSPP-07939 MSPP-07951
BloodStor® 100 Freezing Medium
Cell Freezing Media
Biopreservation reagent for cells and tissues.
  • Sterile, USP vial
  • USP grade/highest-quality components
  • Filled under cGMP
  • Sterility and endotoxin testing
Cryopreserve stem cells and other cells isolated from umbilical cord blood, peripheral blood, bone marrow, and other biologics with BloodStor® 100, an animal component-free and serum-free biopreservation medium. BloodStor® 100 has been sterilized, tested for endotoxin and manufactured under cGMPs with USP-grade components. BloodStor® 100 contains 100% (w/v) DMSO and is available in a variety of formats.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').