CryoStor® CSB Cryopreservation Medium
Supplier: STEMCELL Technologies
100-0239 100-0238
MSPP-100-0239EA 10501.15 USD
MSPP-100-0239 MSPP-100-0238
CryoStor® CSB Cryopreservation Medium
Cell Freezing Media
Animal component-free and protein-free cryopreservation formulation.
  • Serum-free and chemically defined
  • USP grade/multi-compendial/high-quality components
  • cGMP-manufactured
  • Sterility, endotoxin, and cell-based release testing
  • FDA master file
Reduce the level of cryopreservation-induced delayed-onset cell death and maximize post-thaw cell recovery, viability, and function following cryopreservation, with CryoStor® CSB, an intracellular-like base formulation from the CryoStor® preservation media series. Serum-free, animal component-free, and cGMP-manufactured, this chemically defined medium is sterile and tested for endotoxins and cell-based release. CryoStor® CSB is formulated with USP-grade components and does not contain DMSO.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').