Bovine Casein hydrolysate enzymatic (from Milk), MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0210129001 0210129005
IC10129001EA 178.99 USD
IC10129001 IC10129005
Bovine Casein hydrolysate enzymatic (from Milk), MP Biomedicals
Proteins and Peptides
Casein hydrolysate enzymatic is a pancreatic hydrolysate of casein.
  • Creamy Powder
  • Soluble in Water
  • Recommended for use as Animal Diet
  • Ash Content: 0-7.5%
  • Loss on drying: 1-7%
  • Nitrogen Total: ≥11%, Nitrogen (amino): ≥6%
This hydrolysate contains, in the form of mixed amino acids and peptides, all amino acids originally present in casein. It is a refined product providing high quality soluble source of amino acids, polypeptides and peptides.