Human Recombinant Carbonic Anhydrase XII (CA 12) Protein (His and AVI Tag) (from HEK293 Cells), Biotinylated
Supplier: Sino Biological
10617H49HB100 10617H49HB20
77294-698EA 688 USD
77294-698 77294-700
Human Recombinant Carbonic Anhydrase XII (CA 12) Protein (His and AVI Tag) (from HEK293 Cells), Biotinylated
Proteins and Peptides
A DNA sequence encoding the human CA12 (NP_001209.1)(Met1-Gln291) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the biotin-protein ligase (BirA enzyme) which is co-expressed.