Human Recombinant KLRD1 (from HEK293 Cells)
Supplier: Sino Biological
12649-H01H-1 12649-H01H-100
76778-938EA 2704.46 USD
76778-938 76777-830
Human Recombinant KLRD1 (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human KLRD1 (Q13241-1) (Lys32-Ile179) was expressed with the Fc region of human IgG1 at the N-terminus.