Human Recombinant CD19 (from HEK293 Cells) Biotinylated
Supplier: Sino Biological
11880H49HB20 11880H49HB100
76778-796EA 311.41 USD
76778-796 76778-794
Human Recombinant CD19 (from HEK293 Cells) Biotinylated
Proteins and Peptides
A DNA sequence encoding the human CD19 (P15391-1) (Met1-Lys291) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.