Human Recombinant ULBP2 Protein (His & AVI Tag) (from HEK293 Cells), Biotinylated
Supplier: Sino Biological
12143-H27H-B-100 12143-H27H-B-20
76641-374EA 688 USD
76641-374 76642-220
Human Recombinant ULBP2 Protein (His & AVI Tag) (from HEK293 Cells), Biotinylated
Proteins and Peptides
A DNA sequence encoding the human ULBP2 (NP_079493.1) (Met1-Ser217) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.