Human Recombinant IL4/Interleukin-4 Protein (from E. coli), Biotinylated
Supplier: Sino Biological
11846-HNAE-B-20 11846-HNAE-B-100
76642-280EA 248 USD
76642-280 76641-202
Human Recombinant IL4/Interleukin-4 Protein (from E. coli), Biotinylated
Proteins and Peptides
A DNA sequence encoding the IL4 (P05112-1) (His25-Ser153) was expressed with a N-terminal Met. The purified protein was biotinylated in vitro.