Human Recombinant c-Met/HGFR Protein (His & AVI Tag) (from HEK293 Cells), Biotinylated
Supplier: Sino Biological
10692-H27H-B-100 10692-H27H-B-20
76641-474EA 688 USD
76641-474 76642-344
Human Recombinant c-Met/HGFR Protein (His & AVI Tag) (from HEK293 Cells), Biotinylated
Proteins and Peptides
A DNA sequence encoding the human MET (NP_000236.2) (Met1-Thr932) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.