Human Recombinant Glypican 3/GPC3/OCI-5 Protein (His Tag) (from HEK293 Cells), Biotinylated
Supplier: Sino Biological
10088-H08H-B-100 10088-H08H-B-20
76641-392EA 468 USD
76641-392 76641-420
Human Recombinant Glypican 3/GPC3/OCI-5 Protein (His Tag) (from HEK293 Cells), Biotinylated
Proteins and Peptides
A DNA sequence encoding the GPC3 (P51654-1) (Met1-His559) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.