Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)

Supplier: Thermo Scientific
33391-5000
TS33391-5000EA 363 USD
TS33391-5000
Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
Proteins and Peptides
  • Infrared spectrum: Conforms
  • Specific optical rotation: −14 to –18° (20 °C, 589 nm) (C=1, 1 N HCl)
  • EINECS: 261-529-2
  • GHS symbol and signal word: GHS07 and warning
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR