Native Bovine Bovine Serum Albumin (from Serum), Semicarbazide (SEM)
Supplier: AAT Bioquest
50007
76484-028EA 604.2 USD
76484-028
Native Bovine Bovine Serum Albumin (from Serum), Semicarbazide (SEM)
Proteins and Peptides
Semicarbazide (SEM)-BSA Conjugate is used for developing anti-Semicarbazide (SEM) antibodies that are typically used in an ELISA assay.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Caution: For research use only.