Mouse Recombinant IL5Ra/CD125 Protein (His Tag) (from Baculovirus Insect Cells)
Supplier: Sino Biological
51154-M08B-50 51154-M08B-500
103668-692EA 148.35 USD
103668-692 103668-694 76642-086
Mouse Recombinant IL5Ra/CD125 Protein (His Tag) (from Baculovirus Insect Cells)
Proteins and Peptides
A DNA sequence encoding the mouse IL5Ra (P21183) (Met1-Val328) was expressed with a C-terminal polyhistidine tag.