Human Recombinant Tie2/CD202b/TEK Protein (ECD, His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10700-H08H-100 10700-H08H-1
103621-444EA 179.78 USD
103621-444 76777-898
Human Recombinant Tie2/CD202b/TEK Protein (ECD, His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met1-Lys745) of human Tie2 (AAA61139.1) precursor was fused with a polyhistidine tag at the C-terminus.