Mouse Recombinant PD1/PDCD1/CD279 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
50124-M02H-50 50124-M02H-1
103918-636EA 179.78 USD
103918-636 76642-480
Mouse Recombinant PD1/PDCD1/CD279 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse PDCD1 (NP_032824.1) (Met1-Gln167) was expressed with the Fc region of human IgG1 at the C-terminus.