Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 350
Supplier: AAT Bioquest
23110
76483-868EA 180.84 USD
76483-868
Native Amanita phalloides Phalloidin (from Eukaryotic cells), Alexa Fluor® 350
Proteins and Peptides
This blue fluorescent phalloidin conjugate (equivalent to Alexa Fluor® 350-labeled phalloidin) selectively binds to F-actins.
  • Suitable for all fluorescent imaging platforms
  • Specific for actin in both living or fixed cells
  • Multiple applications
Caution: For research use only.