Human Recombinant Noggin/NOG Protein (from HEK293 Cells)
Supplier: Sino Biological
10267-HNAH-1 10267-HNAH-100 10267-HNAH-20
103885-830EA 2161.89 USD
103885-830 103885-834 76641-234
Human Recombinant Noggin/NOG Protein (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human NOG (NP_005441.1) (Met1-Cys232) was expressed.