HardyDisks™ AST Omadacycline 30 µg (OMC30), Nuzyra, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z9411
Z9415
76501-654EA
16.88
USD
76501-654
76501-656
HardyDisks™ AST Omadacycline 30 µg (OMC30), Nuzyra, Hardy Diagnostics
Microbiology Test Systems
Microorganism Tests
HardyDisk™ AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.