N-Boc-glycylglycine

Supplier: Ambeed
A152188-5G A152188-1G A152188-5G A152188-25G A152188-100G A152188-10G A152188-1G
103842-516EA 6 USD
103842-516 77318-130 77299-930 77095-854 77567-196 103804-356 103856-878
N-Boc-glycylglycine
Proteins and Peptides
  • Boiling point: 488 °C (760 mm Hg)
  • GHS symbol: GHS07
  • Signal word: Warning

For Research Use Only

Caution: Sealed in dry, store in freezer, under –20 °C.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR