Carbonylcyanide-3-chlorophenylhydrazone ≥98%

Supplier: Cayman Chemical Company

Synonyms: CCCP, Carbonyl cyanide m-chlorophenylhydrazone, Carbonyl cyanide m-chlorophenylhydrazone

25458-250 25458-500 25458-100
76304-452EA 89.02 USD
76304-452 76304-454 76304-462
Carbonylcyanide-3-chlorophenylhydrazone ≥98%
Carbonylcyanide-3-chlorophenylhydrazone

CCCP inhibits mitochondrial respiration and ATPase activity when used at concentrations of 110 and 35 nM, respectively; inhibits activation of STING at 50 µM; increases body temperature as well as left ventricular systolic and diastolic dimensions and decreases fractional shortening in rats at 4 mg/kg


  • A mitochondrial uncoupler
  • Crystalline solid
  • Stability: ≥ 2 years

Formula: C₉H₅ClN₄
MW: 204.617 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00001848
CAS Number: 555-60-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77030e0d7a68562df21f2cf0d05a54f34e0f1266&vid=77030e0d7a68562df21f2cf0d05a54f34e0f1266&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR