Human Recombinant ADAM5P
Supplier: Abnova
Human ADAM5P full-length ORF (AAH67864.2, 1 a.a. to 412 a.a.) recombinant protein with GST-tag at N-terminal.
- Host: Wheat germ (in vitro)
- Preparation method: in vitro wheat germ expression system
- Storage buffer: 50 mM Tris-HCI, 10 mM reduced glutathione, pH=8.0 in the elution buffer
- Gene description: ADAM metallopeptidase domain 5 pseudogene
Applications include enzyme-linked immunoabsorbent assay, western blot (recombinant protein), antibody production and protein array.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...