Human HLA-DRB1*01:01 raw Monomer
Supplier: MBL International
TBCM4-R1011-RM
103500-482EA 13846.15 USD
103500-482
Human HLA-DRB1*01:01 raw Monomer
Proteins and Peptides
MHC Tetramers are used for the detection of antigen-specific T-cell populations.
  • Class-II
MHC-peptide tetramer complexes recognize antigen specific T-cells at the single cell level, allowing for accurate measurements of the functional immune response in cancer, infectious diseases, and autoimmune diseases. Due to their increased avidity, MHC tetramers stably bind to cognate TcRs and are evaluated using flow cytometry.