Mouse H-2Dk, Brilliant Violet® 421
Supplier: MBL International
TBCM3-DKI-4
103500-454EA 3076.92 USD
103500-454
Mouse H-2Dk, Brilliant Violet® 421
Proteins and Peptides
MHC Tetramers are used for the detection of antigen-specific T-cell populations.
  • Class-I
MHC-peptide tetramer complexes recognize antigen specific T-cells at the single cell level, allowing for accurate measurements of the functional immune response in cancer, infectious diseases, and autoimmune diseases. Due to their increased avidity, MHC tetramers stably bind to cognate TcRs and are evaluated using flow cytometry.