Human HLA-A*02:01, Biotin

Supplier: MBL International
TB-0154-M
103500-338EA 2153.85 USD
103500-338
Human HLA-A*02:01, Biotin
Proteins and Peptides
MHC Tetramers for detecting antigen specific t-cell populations.

  • Class I
  • Application: FCM

Used in immunology and immune monitoring.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR