Human HLA-B*27:05, Biotin

Supplier: MBL INTERNATIONAL CORP MS
TB-0148-M
103500-512EA 2153.85 USD
103500-512
Human HLA-B*27:05, Biotin
Proteins and Peptides
MHC Tetramers for detecting antigen specific t-cell populations.

  • Class I
  • Application: FCM

Used in immunology and immune monitoring.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR