Human HLA-A*01:01

Supplier: MBL International
TB-0125-2
103500-246EA 2307.69 USD
103500-246
Human HLA-A*01:01
Proteins and Peptides
MHC Tetramers are used for the detection of antigen-specific T-cell populations.

  • Class I
  • Application: FCM

MHC-peptide tetramer complexes recognize antigen specific T-cells at the single cell level, allowing for accurate measurements of the functional immune response in cancer, infectious diseases, and autoimmune diseases.  Due to their increased avidity, MHC tetramers stably bind to cognate TcRs and are evaluated using flow cytometry.

Used in immunology and immune monitoring.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR