Rabbit Plasma Sterile In ACD, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D509-01-0100 D509-01-0050
75935-116EA 1272.54 USD
75935-116 75935-118
Rabbit Plasma Sterile In ACD, Rockland Immunochemicals
Blood
Sterile Plasma in Anticoagulant
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by collection of whole blood into said anticoagulant
  • Red blood cells are removed through a process of centrifugation and filtration techniques - leaving non-sterile plasma behind
used as a component of bioassays, immunoassays or enzyme assays
used as a supplement to cell culture media
provides a broad spectrum of macromolecules, carrier proteins for lipoid substances andMore Product Information