Sheep Plasma Non Sterile In Lithium Heparin, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D411-07-1000 D411-07-0100 D411-07-0050 D411-07-0500
75935-272EA 603.88 USD
75935-272 75935-274 75935-276 75935-278
Sheep Plasma Non Sterile In Lithium Heparin, Rockland Immunochemicals
Blood
Non-Sterile Plasma in Anticoagulant
  • prepared by collection of whole blood into said anticoagulant
  • Red blood cells are removed through a process of centrifugation and filtration techniques - leaving non-sterile plasma behind
used as a component of bioassays, immunoassays or enzyme assays
provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health
also available as a sterileMore Product Information