Guinea Pig Plasma Non-Sterile In Potassium Oxalate, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D405-08-0100 D405-08-0050
75928-324EA 1501.34 USD
75928-324 75928-326
Guinea Pig Plasma Non-Sterile In Potassium Oxalate, Rockland Immunochemicals
Blood
Non-Sterile Plasma in Anticoagulant
  • prepared by collection of whole blood into said anticoagulant
  • Red blood cells are removed through a process of centrifugation and filtration techniques - leaving non-sterile plasma behind
used as a component of bioassays, immunoassays or enzyme assays
provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health
also available as a sterileMore Product Information