Goat Plasma Non-Sterile In ACD, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D404-01-1000 D404-01-0100 D404-01-0050 D404-01-0500
10801-804EA 603.88 USD
10801-804 10801-806 10802-984 10802-986
Goat Plasma Non-Sterile In ACD, Rockland Immunochemicals
Blood
Non-Sterile Plasma in Anticoagulant
  • prepared by collection of whole blood into said anticoagulant
  • Red blood cells are removed through a process of centrifugation and filtration techniques - leaving non-sterile plasma behind
used as a component of bioassays, immunoassays or enzyme assays
provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health
also available as a sterile preparation for use as a cell culture supplement
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal