Rat Serum Non Sterile, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D210-00-0100 D210-00-0050
75935-196EA 994.63 USD
75935-196 75935-200
Rat Serum Non Sterile, Rockland Immunochemicals
Sera (Serum)
Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Rat Serum is used as a component of bioassays, immunoassays or enzyme assays. Rat Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Rat serum is also available as a sterile preparation for use as a cell culture supplement.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal