GUINEA PIG SERUM (non-sterile), Rockland Immunochemicals
Supplier: Rockland Immunochemical
D205-00-0050 D205-00-0100
75928-664EA 840 USD
75928-664 75928-666
GUINEA PIG SERUM (non-sterile), Rockland Immunochemicals
Sera (Serum)
Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Guinea Pig Serum is used as a component of bioassays, immunoassays or enzyme assays. Guinea Pig Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Guinea Pig Serum is also available as a sterile preparation for use as a cell culture supplement. Guinea Pig Serum is ideal for investigators in Cancer, Immunology and Cell Biology research.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal