DONKEY SERUM (sterile), Rockland Immunochemicals
Supplier: Rockland Immunochemical
D120-00-0050 D120-00-0100
RLD120-00-0050EA 484.78 USD
RLD120-00-0050 RLD120-00-0100
DONKEY SERUM (sterile), Rockland Immunochemicals
Sera (Serum)
Sterile Serum
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Donkey Serum is used as a supplement to cell culture media. Donkey Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, andMore Product Information