GOAT SERUM (sterile), Rockland Immunochemicals
Supplier: Rockland Immunochemical
D104-00-0500 D104-00-0100 D104-00-1000
75928-600EA 548 USD
75928-600 75934-358 75934-360
GOAT SERUM (sterile), Rockland Immunochemicals
Sera (Serum)
Sterile Serum
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Goat Serum is used as a supplement to cell culture media. Goat Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, andMore Product Information