Mouse Thrombopoietin
Supplier: Rockland Immunochemicals
010-001-W32-0010 010-001-W32-0002 010-001-W32-0100 010-001-W32-1000
75932-884EA 961.19 USD
75932-884 75932-876 75932-888 75932-880
Mouse Thrombopoietin
Proteins and Peptides
Recombinant Mouse Thrombopoietin control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Thrombopoietin (TPO) is a growth factor that is produced by the liver and kidney. TPO acts through the TPO receptor to promote megakaryocyte maturation and differentiation, which leads to the production of platelets. Recombinant mouse TPO is a non-glycosylated protein, containing 174 amino acids (which comprise the receptor binding domain), with a molecular weight of 18.7 kDa.
Thrombopoietin Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Thrombopoietin in immunological assays.