Mouse Recombinant IL-5
Supplier: Rockland Immunochemicals
010-001-V42-1000
75929-174EA 4582 USD
75929-174
Mouse Recombinant IL-5
Proteins and Peptides
Recombinant Mouse IL-5 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-5 (IL-5) is a hematopoietic growth factor expressed in Th2, mast cells and eosinophils. IL-5 acts through its receptor, the IL-5 receptor (IL-5R) and is involved in B cell growth and eosinophil activation. IL-5 has been shown to be and is regulated by GATA-3, in addition to other transcription factors. Human and mouse IL-5 are cross-reactive. Recombinant mouse IL-5 is a non-glycosylated, disulfide-linked homodimer, containing two 113 amino acid chains, with a total molecular weight of 26.2 kDa.
Interleukin-5 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-5 in immunological assays.