TAF1 (N-Term)
Supplier: Rockland Immunochemicals
009-001-T71S 009-001-T71
75932-562EA 714 USD
75932-562 75932-560
TAF1 (N-Term)
Proteins and Peptides
TAF1 (N-term) control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
TAF1 is a DNA-binding protein complex required for RNA polymerase II-mediated transcription of many, if not all, protein-encoding genes in eukaryotic cells. TAF1 plays a key role in the initiation process, since it binds to the TATA element to form a complex that nucleates the assembly of the other components into a preinitiation complex and that may be stable through multiple rounds of transcription. TAF1 may be targeted to specific chromatin-bound promoters and may play a key role in chromatin recognition (1). TAF1 also serves to link the control of transcription to the cell cycle (2). TAF1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cell Cycle, and Invasion/Metastasis research.
TAF1 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. TAF1 Protein is suitable for use in Western Blot. Expect a band approximately ~130kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.