PSKH1
Supplier: Rockland Immunochemicals
009-001-T07S 009-001-T07
75935-048EA 714 USD
75935-048 75935-046
PSKH1
Proteins and Peptides
PSKH1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PSKH1 is a member of the family of protein serine kinase H1 which was originally cloned by homology probing (1). PSKH1 has been localized to the endoplasmic reticulum (ER), the Golgi apparatus, and the plasma membrane (PM). PSKH1 has been shown to play a structural and regulatory role in maintenance of the Golgi apparatus, which is a key organelle within the secretory pathway. PSKH1 is in a cluster of 5 unrelated genes on 16q22.1 chromosome which is transcribed in the opposite direction from that of LCAT which is a member of the same cluster (2). PSKH1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cardiovascular Disease, ERK/MAPK Pathway, Neurobiology, PKA/PKC Pathway, and Ser/Thr Kinases research.
PSKH1 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PSKH1 Protein is suitable for use in Western Blot. Expect a band approximately ~72kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.