ING2
Supplier: Rockland Immunochemicals
009-001-S65S 009-001-S65
75930-698EA 714 USD
75930-698 75930-696
ING2
Proteins and Peptides
p33ING2 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
p33ING2 is a member of the inhibitor of growth (ING) family that is associate with and modulates the activity of histone acetyltransferase (HAT) and histone deacetylase (HDAC) complexes and function in DNA repair and apoptosis. P33ING2 is a DNA damage-inducible gene that negatively regulates cell proliferation through activation of p53 by enhancing its acetylation (1). P33ING2 is required for initial DNA damage sensing and chromatin remodeling in the nucleotide excision repair process (2). p33ING2 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, and Cell Cycle research.
p33ING2 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. p33ING2 Protein is suitable for use in Western Blot. Expect a band approximately ~62kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.