HSP90 alpha control protein-HIS Epitope
- Control Protein
- PCA Assays
- ImmunoBlot
HSP90α is a member of the HSP90 family of proteins which are important molecular chaperones involved in signal transduction, cell cycle control, stress management, folding, degradation, and transport of proteins (1). HSP90 is a molecular chaperone that plays a key role in the conformational maturation of oncogenic signaling proteins, including HER2/ERBB2, AKT, RAF1, BCR-ABL and mutated p53. HSP90 inhibitors bind to HSP90, and induce the proteasomal degradation of HSP90 client proteins. HSP90α is an important mediator of cancer cell invasion and is expressed extracellularly on fibrosarcoma and breast cancer cells where it interacts with MMP2 (2). HSP90α Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, AKT/PKB Pathway, Apoptosis/Autophagy, Cancer, and Cellular Stress research.
HSP90α Protein is stored in 50mM MOPS, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. HSP90α Protein is suitable for use in Western Blot. Expect a band approximately ~94kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.