VIM
Supplier: Rockland Immunochemicals
009-001-E54S 009-001-E54
75933-558EA 364 USD
75933-558 75933-560
VIM
Proteins and Peptides
Vimentin control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Vimentin is a member of intermediate filament family of proteins and is an important structural feature of eukaryotic cells (1). Along with microtubules and actin microfilaments, Vimentin make up the cytoskeleton component of cells. Studies have shown that Vimentin is attached to the nucleus, endoplasmic reticulum and mitochondria, either laterally or terminally. Vimentin plays a significant role in supporting and anchoring the position of the organelles in the cytosol (2). Thus, Vimentin plays a key role in maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. Vimentin Protein is ideal for investigators involved in Signaling Proteins, Microtubule/Actin Associated Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cell Cycle, Cellular Stress, Inflammation, Invasion/Metastasis, and Neurobiology research.
Vimentin Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. Vimentin Protein is suitable for use in Western Blot. Expect a band approximately ~60kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.