Rat Recombinant csf2 (from E. coli)
Supplier: Rockland Immunochemicals
012-001-V12-0005 012-001-V12-1000
75934-450EA 154.63 USD
75934-450 75928-204
Rat Recombinant csf2 (from E. coli)
Proteins and Peptides
Granulocyte Macrophage Colony Stimulating Factor (GM-CSF) is hematopoietic factor produced by entothelial cells, monocytes, fibroblasts and T cells in response to a number of inflammatory mediators. GM-CSF is able to stimulate the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells. GM-CSF can also stimulate some functional activities in mature granulocytes and macrophages. Human and mouse GM-CSF show no cross-reactivity. Recombinant rat GM-CSF is a non-glycosylated protein, containing 128 amino acids, with a molecular weight of 14.6 kDa.