Human Recombinant EPHA8 (from Baculovirus (Sf9 Insect cells))
Supplier: Rockland Immunochemicals
009-001-R90 009-001-R90S
10801-238EA 714 USD
10801-238 10801-236
Human Recombinant EPHA8 (from Baculovirus (Sf9 Insect cells))
Proteins and Peptides
EPHA8 is a member of the ephrin receptor subfamily of the protein-tyrosine kinase family in ahich EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats (1). EPHA8 receptors play a role in axonal pathfinding during development of the mammalian nervous system (2). EPHA8 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Angiogenesis, Cancer, Cardiovascular Disease, Neurobiology, and Receptor Tyrosine Kinases research.