Trio.Bas™ DUO ATEX Kits, Hardy Diagnostics
Supplier: Hardy Diagnostics
BAS245K BAS246K BAS247K BAS248K
76288-324EA 20568 USD
76288-324 76288-326 76288-328 76288-352
Trio.Bas™ DUO ATEX Kits, Hardy Diagnostics
Samplers Air Samplers
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
  • Two aspirating heads with easy manipulation bayonet closure
  • Ability to use two different culture media at the same time
  • Save sampling time by doubling the aspirated volume of air
  • Manual or automatic operations
The Trio.Bas™ instruments utilize the impaction method of air sampling. The air, passing through an aspirating head, impacts onto an appropriate agar plate that collects bacteria or other organic substances possibly present in the sampled area.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable