Mouse Recombinant IL17RA/IL17R/CD217 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
50328-M02H-100 50328-M02H-1
103664-900EA 300.98 USD
103664-900 103664-902 76641-950
Mouse Recombinant IL17RA/IL17R/CD217 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mouse IL17RA (Q60943)(Met1-Trp322) was expressed with the Fc region of human IgG1 at the C-terminus.