Mouse Recombinant Oncostatin M/OSM Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
50112-M08H-100 50112-M08H-1
103664-604EA 351.48 USD
103664-604 103666-788 103677-922 76641-450
Mouse Recombinant Oncostatin M/OSM Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mature form of mouse OSM (P53347) (Met 1-Arg 206) was expressed, with a C-terminal polyhistidine tag.